Chemical Identifiers
CAS531-75-9
IUPAC Name7-hydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
Molecular FormulaC15H16O9
InChI KeyXHCADAYNFIFUHF-TYKRLAFXNA-N
SMILESOC[C@H]1O[C@@H](OC2=C(O)C=C3OC(=O)C=CC3=C2)[C@H](O)[C@@H](O)[C@@H]1O
View more