Gateway™ BP Clonase™ II enzyme mix catalyzes the in vitro recombination of PCR products or subcloning DNA segments from clones (containing attB sites) and a donor vector (containing attP sites) to generate entry clones. Gateway™ BP Clonase™ II contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set up with fewer pipetting steps.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Shipping Condition
Dry Ice
Enzyme
BP Clonase
Compatible Buffer
Enzyme Buffer
Quantity
20 Reactions
Product Type
BP Clonase Enzyme Mix
Product Line
Clonase™, Gateway™
Unit Size
20 reactions
Contents & Storage
Gateway™ BP Clonase™ II enzyme mix includes proteinase K solution (2 μg/μl) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored.