Thermo Scientific Chemicals

2-Ethoxyethyl Ether, 98+%, Extra Pure, Thermo Scientific Chemicals

Catalog number: 117905000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Ethoxyethyl Ether, 98+%, Extra Pure, Thermo Scientific Chemicals

Catalog number: 117905000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 117905000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
112-36-7
IUPAC Name
1-ethoxy-2-(2-ethoxyethoxy)ethane
Molecular Formula
C8H18O3
InChI Key
RRQYJINTUHWNHW-UHFFFAOYSA-N
SMILES
CCOCCOCCOCC
Infrared spectrum
Conforms
Acidity (CH3COOH)
=<0.015 %
Appearance (Form)
Clear liquid
GC
>=98.0 %
Residue after evaporation
=<0.05 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text