Invitrogen™
Gateway™ LR Clonase™ II Enzyme mix
Catalog number: 11791100
Have Questions?
Invitrogen™
Gateway™ LR Clonase™ II Enzyme mix
Catalog number: 11791100
Have Questions?
Catalog Number
11791100
Unit Size
100 rxns
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
20 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
11791100100 rxns
-
1179102020 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Gateway™ LR Clonase™ II enzyme mix catalyzes the in vitro recombination between an entry clone (containing a gene of interest flanked by attL sites) and a destination vector (containing attR sites) to generate an expression clone. Gateway™ LR Clonase™ II contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set-up with fewer pipetting steps.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Shipping Condition
Dry Ice
Enzyme
LR Clonase
Compatible Buffer
Enzyme Buffer
Quantity
100 Reactions
Product Type
LR Clonase Enzyme Mix
Product Line
Clonase™, Gateway™
Contents & Storage
Gateway™ LR Clonase™ II enzyme mix includes proteinase K solution (2 μg/μL) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored.