EXPRESS SYBR™ GreenER™ qPCR Supermix, with premixed ROX
Product Image
Invitrogen™

EXPRESS SYBR™ GreenER™ qPCR Supermix, with premixed ROX

Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
1179401K
also known as 11794-01K
1000 Reactions
11794200200 Reactions
2 Options
Catalog number 1179401K
also known as 11794-01K
Price (USD)/ 1,000 reactions
3,245.00
-
Add to cart
No. of Reactions:
1000 Reactions
Price (USD)/ 1,000 reactions
3,245.00
Add to cart
EXPRESS SYBR™ GreenER™ qPCR Supermix, with premixed ROX
Catalog number1179401K
Price (USD)/ 1,000 reactions
3,245.00
-
Add to cart
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of EXPRESS SYBR GreenER SuperMix and added additional capabilities for your gene expression analysis.

EXPRESS SYBR GreenER qPCR SuperMix kits are compatible with fast-cycling instruments for your SYBR-based qPCR experiments. This EXPRESS SYBR GreenER qPCR SuperMix comes with premixed ROX reference dye. EXPRESS SYBR GreenER qPCR Supermix Universal, with separate ROX reference dye, is also available.

This kit provides:
• Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
• SYBR GreenER dye
• UDG carryover protection—uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodSYBR
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
PolymeraseTaq DNA Polymerase
Reaction SpeedFast or Standard
For Use With (Equipment)7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
Kit TypeFast-Cycling Instrument Kit
Product LineSYBR™ GreenER™
Product TypeReal Time PCR SYBR Master Mix
Sufficient For1000 Reactions
For Use With (Application)Gene Expression
No. of Reactions1000 Reactions
Sample TypeDNA (Genomic), cDNA
Volume5 x 5 mL
Unit Size1,000 reactions
Contents & Storage
Contains 5 × 5 ml EXPRESS SYBR™ GreenER™ qPCR SuperMix with Premixed ROX. Store at −20°C for long-term storage. EXPRESS qPCR SuperMixes may be stored at 4–8°C for up to one month.