Invitrogen™

EXPRESS qPCR Supermix, with premixed ROX

Catalog number: 1179501K
Have Questions?
Invitrogen™

EXPRESS qPCR Supermix, with premixed ROX

Catalog number: 1179501K
Have Questions?
Catalog Number
1179501K
also known as 11795-01K
Unit Size
1,000 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1179501K
also known as 11795-01K
1,000 reactions
-
11795200200 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of EXPRESS qPCR SuperMix and added additional capabilities for your gene expression analysis.

EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for your probe-based qPCR assays. This kit comes with premixed ROX reference dye. EXPRESS qPCR SuperMix kits with a separate tube of ROX reference dye are also available.

This kit provides:
• Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
• UDG carryover protection—uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
Taq DNA Polymerase
Reaction Speed
Fast or Standard
For Use With (Equipment)
7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
Product Type
Express qPCR Supermix
Sufficient For
1000 Reactions
For Use With (Application)
Gene Expression
No. of Reactions
1000 Reactions
Sample Type
DNA (Genomic), cDNA

Contents & Storage

EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube.

Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text