Thermo Scientific Chemicals

Ethyl acetoacetate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 117970250
25 L, Plastic drum, Each
Thermo Scientific Chemicals

Ethyl acetoacetate, 99%, pure, Thermo Scientific Chemicals

Catalog number: 117970250
25 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 117970250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
141-97-9
IUPAC Name
ethyl 3-oxobutanoate
Molecular Formula
C6H10O3
InChI Key
XYIBRDXRRQCHLP-UHFFFAOYSA-N
SMILES
CCOC(=O)CC(C)=O
GC
>=98.5 %
Refractive index
1.4180 to 1.4200 (20°C, 589 nm)
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text