Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
25 mL
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 117980010
Price (USD)/ Each
60.65
Online exclusive
71.00 
Save 10.35 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
60.65
Online exclusive
71.00 
Save 10.35 (15%)
Add to cart
Ethyl acrylate, 99.5%, stabilized, Thermo Scientific Chemicals
Catalog number117980010
Price (USD)/ Each
60.65
Online exclusive
71.00 
Save 10.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameethyl prop-2-enoate
Molecular FormulaC5H8O2
InChI KeyJIGUQPWFLRLWPJ-UHFFFAOYSA-N
SMILESCCOC(=O)C=C
Molecular Weight (g/mol)100.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Stabilizer10 to 20 ppm MEHQ
GC>=99.40 %
Refractive index1.4050 to 1.4070 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only