Thermo Scientific Chemicals

Ethyl 3-aminobenzoate, 99+%, Thermo Scientific Chemicals

Catalog number: 117990050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 3-aminobenzoate, 99+%, Thermo Scientific Chemicals

Catalog number: 117990050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 117990050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
582-33-2
IUPAC Name
ethyl 3-aminobenzoate
Molecular Formula
C9H11NO2
InChI Key
ZMCBYSBVJIMENC-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=CC(N)=C1
Appearance (Form)
Liquid or low melting solid
Appearance (Color)
Clear colorless to yellow to red-brown to brown
GC
>=99.0 %
Infrared spectrum
Conforms
Refractive index
1.5595 to 1.5635 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text