Appearance (Color)Clear colorless to yellow to red-brown to brown
GC>=99.0 %
Infrared spectrumConforms
Refractive index1.5595 to 1.5635 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.