Ethyl 3-aminobenzoate, methanesulfonic acid salt, 98%, Thermo Scientific Chemicals
Ethyl 3-aminobenzoate, methanesulfonic acid salt, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3-aminobenzoate, methanesulfonic acid salt, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
250 g
Packaging:
View all
Plastic bottle
Catalog number 118000500
Price (USD)/ Each
160.65
Online exclusive
178.00 
Save 17.35 (10%)
-
Add to cart
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
160.65
Online exclusive
178.00 
Save 17.35 (10%)
Add to cart
Ethyl 3-aminobenzoate, methanesulfonic acid salt, 98%, Thermo Scientific Chemicals
Catalog number118000500
Price (USD)/ Each
160.65
Online exclusive
178.00 
Save 17.35 (10%)
-
Add to cart
Chemical Identifiers
CAS886-86-2
IUPAC Nameethyl 3-aminobenzoate; methanesulfonic acid
Molecular FormulaC10H15NO5S
InChI KeyFQZJYWMRQDKBQN-UHFFFAOYSA-N
SMILESCS(O)(=O)=O.CCOC(=O)C1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point146°C to 150°C
Appearance (Form)Powder
Solubility(5% in Water) Clear solution
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only