Thermo Scientific Chemicals

Ethyl 3-aminobenzoate, methanesulfonic acid salt, 98%, Thermo Scientific Chemicals

Catalog number: 118002500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ethyl 3-aminobenzoate, methanesulfonic acid salt, 98%, Thermo Scientific Chemicals

Catalog number: 118002500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 118002500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
886-86-2
IUPAC Name
ethyl 3-aminobenzoate; methanesulfonic acid
Molecular Formula
C10H15NO5S
InChI Key
FQZJYWMRQDKBQN-UHFFFAOYSA-N
SMILES
CS(O)(=O)=O.CCOC(=O)C1=CC=CC(N)=C1
Appearance (Color)
White to off-white
Melting point
146°C to 150°C
Appearance (Form)
Powder
Solubility
(5% in Water) Clear solution
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text