Search Thermo Fisher Scientific
Catalog Number | For Use With (Equipment) |
---|---|
118085 | SWB1122, RSWB3222 |
118081 | Water Bath Models WB1110 |
118082 | Water Bath Models WB1120 |
118083 | Water Bath Models WB1130 |
118084 | WB1140 |
Minimize heat loss and maintain bath temperature until ready for next application with Thermo Scientific™ Flat Stainless-Steel Covers.