Thermo Scientific Chemicals

Ethyl 4-bromocrotonate, 75%, tech., Thermo Scientific Chemicals

Catalog number: 118121000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 4-bromocrotonate, 75%, tech., Thermo Scientific Chemicals

Catalog number: 118121000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118121000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
37746-78-4
IUPAC Name
ethyl (2E)-4-bromobut-2-enoate
Molecular Formula
C6H9BrO2
InChI Key
FHGRPBSDPBRTLS-ONEGZZNKSA-N
SMILES
CCOC(=O)\C=C\CBr
Appearance (Color)
Clear light yellow to brown
Infrared spectrum
Conforms
Appearance (Form)
Liquid
GC
>=70 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text