Thermo Scientific Chemicals

Ethyl 2-bromoisobutyrate, 98%, Thermo Scientific Chemicals

Catalog number: 118130050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 2-bromoisobutyrate, 98%, Thermo Scientific Chemicals

Catalog number: 118130050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118130050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
600-00-0
IUPAC Name
ethyl 2-bromo-2-methylpropanoate
Molecular Formula
C6H11BrO2
InChI Key
IOLQWGVDEFWYNP-UHFFFAOYSA-N
SMILES
CCOC(=O)C(C)(C)Br
Appearance (Form)
Liquid
GC
>=97.5 %
Appearance (Color)
Colorless to yellow
Infrared spectrum
Conforms
Refractive index
1.4430 to 1.4460 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text