Thermo Scientific Chemicals

DL-Ethyl 2-bromopropionate, 99%, Thermo Scientific Chemicals

Catalog number: 118142500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-Ethyl 2-bromopropionate, 99%, Thermo Scientific Chemicals

Catalog number: 118142500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118142500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
535-11-5
IUPAC Name
ethyl 2-bromopropanoate
Molecular Formula
C5H9BrO2
InChI Key
ARFLASKVLJTEJD-UHFFFAOYNA-N
SMILES
CCOC(=O)C(C)Br
Appearance (Color)
Clear colorless to light yellow
GC
>=98.5 %
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Refractive index
1.4450 to 1.4470 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text