Thermo Scientific Chemicals

Ethyl carbazate, 97%, Thermo Scientific Chemicals

Catalog number: 118201000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl carbazate, 97%, Thermo Scientific Chemicals

Catalog number: 118201000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118201000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4114-31-2
IUPAC Name
ethoxycarbohydrazide
Molecular Formula
C3H8N2O2
InChI Key
VYSYZMNJHYOXGN-UHFFFAOYSA-N
SMILES
CCOC(=O)NN
Appearance (Color)
May have a pink hue
Infrared spectrum
Conforms
Melting point
42°C to 48°C
Appearance (Color)
White to almost white
Appearance (Form)
Crystals and/or chunks or fused solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text