Thermo Scientific Chemicals

Ethyl chloroacetate, 99%, Thermo Scientific Chemicals

Catalog number: 118220100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

Ethyl chloroacetate, 99%, Thermo Scientific Chemicals

Catalog number: 118220100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 118220100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
105-39-5
IUPAC Name
ethyl 2-chloroacetate
Molecular Formula
C4H7ClO2
InChI Key
VEUUMBGHMNQHGO-UHFFFAOYSA-N
SMILES
CCOC(=O)CCl
Infrared spectrum
Conforms
Refractive index
1.4200 to 1.4220 (20°C, 589 nm)
Appearance (Color)
Clear colorless
GC
>=98.5 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text