Ethyl chloroacetate, 99%, Thermo Scientific Chemicals
Ethyl chloroacetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl chloroacetate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 118222500
Price (USD)/ Each
26.90
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
26.90
Add to cart
Ethyl chloroacetate, 99%, Thermo Scientific Chemicals
Catalog number118222500
Price (USD)/ Each
26.90
-
Add to cart
Chemical Identifiers
CAS105-39-5
IUPAC Nameethyl 2-chloroacetate
Molecular FormulaC4H7ClO2
InChI KeyVEUUMBGHMNQHGO-UHFFFAOYSA-N
SMILESCCOC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4200 to 1.4220 (20°C, 589 nm)
Appearance (Color)Clear colorless
GC>=98.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only