Ethyl 3-chloropropionate, 98%, Thermo Scientific Chemicals
Ethyl 3-chloropropionate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3-chloropropionate, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
100 g
Packaging:
Glass bottle
Catalog number 118261000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS623-71-2
IUPAC Nameethyl 3-chloropropanoate
Molecular FormulaC5H9ClO2
InChI KeyZCLGVXACCAZJOX-UHFFFAOYSA-N
SMILESCCOC(=O)CCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4240 to 1.4260 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only