The Baculovirus Expression System with Gateway™ Technology is designed to create recombinant pFastBac™ plasmids containing the polyhedrin promoter. The Baculovirus Expression System with Gateway™ Technology offers:
• Expression using the Bac-to-Bac™ technology (1) • Destination vectors carrying the polyhedrin promoter for production of native (pDEST™8), N-terminal histidine fusion (pDEST™10), and N-terminal GST fusion (pDEST™20) proteins • pDEST™10 vector containing a TEV protease cleavage site for removal of the fusion tag after protein purification (2)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Promoter
Polyhedrin
Product Type
Expression System
Protein Tag
GST Tag, His Tag (6x), Untagged
Cloning Method
Gateway
Vector
pDEST, Gateway Baculovirus Vectors
Product Line
Gateway™
Contents & Storage
The Baculovirus Expression System with Gateway™ Technology includes LR Clonase™ enzyme mix and reaction buffer, destination vectors pDEST™8, pDEST™10, and pDEST™20, proteinase K solution (2 μg/μl), pENTR™-gus positive control, and Library Efficiency™ DH5Competent Cells. The kit does not include MAX Efficiency™ DH10Bac™ Competent Cells. Store LR Clonase™ enzyme mix and Competent Cells at -80°C. Store all other components at -20°C. Components are guaranteed stable for 6 months when properly stored.