Thermo Scientific Chemicals

Ethyl cyanoacetate, 98+%, Thermo Scientific Chemicals

Catalog number: 118280010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl cyanoacetate, 98+%, Thermo Scientific Chemicals

Catalog number: 118280010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 118280010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
105-56-6
IUPAC Name
ethyl 2-cyanoacetate
Molecular Formula
C5H7NO2
InChI Key
ZIUSEGSNTOUIPT-UHFFFAOYSA-N
SMILES
CCOC(=O)CC#N
GC
>=98.0 %
Color scale
=<50 APHA
Water
=<0.1 % (K.F.)
Free acid
=<10 mg/kg (as cyanoacetic acid)
Appearance (Form)
Clear liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text