Ethyl cyanoacetate, 98+%, Thermo Scientific Chemicals
Ethyl cyanoacetate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl cyanoacetate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 118280051
Price (USD)/ Each
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Chemical Identifiers
CAS105-56-6
IUPAC Nameethyl 2-cyanoacetate
Molecular FormulaC5H7NO2
InChI KeyZIUSEGSNTOUIPT-UHFFFAOYSA-N
SMILESCCOC(=O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Color scale=<50 APHA
Water=<0.1 % (K.F.)
Free acid=<10 mg/kg (as cyanoacetic acid)
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only