1x1 image pixel for data collection
Ethyl cyanoacetate, 98+%
Ethyl cyanoacetate, 98+%
Ethyl cyanoacetate, 98+%
Ethyl cyanoacetate, 98+%
Thermo Scientific Chemicals

Ethyl cyanoacetate, 98+%

CAS: 105-56-6 | C5H7NO2 | 113.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 kg
Packaging:
View all
Glass bottle
Catalog number 118282500
Price (USD)
48.65
Online Exclusive
53.80
Save 5.15 (10%)
Each
In stock
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
48.65
Online Exclusive
53.80
Save 5.15 (10%)
Each
Add to cart
Chemical Identifiers
CAS105-56-6
IUPAC Nameethyl 2-cyanoacetate
Molecular FormulaC5H7NO2
InChI KeyZIUSEGSNTOUIPT-UHFFFAOYSA-N
SMILESCCOC(=O)CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid=<10 mg/kg (as cyanoacetic acid)
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.4170 to 1.4190 (20°C, 589 nm)
GC>=98.0 %
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0443337Certificate of AnalysisJun 19, 2025118280010, 118280051, 118282500
A0472803Certificate of AnalysisJun 06, 2025118280010, 118280051, 118282500
A0443337Certificate of AnalysisAug 25, 2023118280010, 118280051, 118282500
A0412192Certificate of AnalysisDec 13, 2022118280010, 118280051, 118282500
A0441917Certificate of AnalysisMay 16, 2022118280010, 118280051, 118282500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection