Ethyl 2-oxocyclohexanecarboxylate, 95%, Thermo Scientific Chemicals
Ethyl 2-oxocyclohexanecarboxylate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-oxocyclohexanecarboxylate, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 118330250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1655-07-8
IUPAC Nameethyl 2-oxocyclohexane-1-carboxylate
Molecular FormulaC9H14O3
InChI KeyFGSGHBPKHFDJOP-UHFFFAOYNA-N
SMILESCCOC(=O)C1CCCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Refractive index1.4775 to 1.4795 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=94 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only