Thermo Scientific Chemicals

Ethyl 2-oxocyclohexanecarboxylate, 95%, Thermo Scientific Chemicals

Catalog number: 118331000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 2-oxocyclohexanecarboxylate, 95%, Thermo Scientific Chemicals

Catalog number: 118331000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118331000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1655-07-8
IUPAC Name
ethyl 2-oxocyclohexane-1-carboxylate
Molecular Formula
C9H14O3
InChI Key
FGSGHBPKHFDJOP-UHFFFAOYNA-N
SMILES
CCOC(=O)C1CCCCC1=O
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4775 to 1.4795 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=94 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text