Thermo Scientific Chemicals

Ethyl dichlorophosphate, 97%, Thermo Scientific Chemicals

Catalog number: 118380250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl dichlorophosphate, 97%, Thermo Scientific Chemicals

Catalog number: 118380250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118380250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1498-51-7
IUPAC Name
ethyl chlorophosphonochloridate
Molecular Formula
C2H5Cl2O2P
InChI Key
YZBOZNXACBQJHI-UHFFFAOYSA-N
SMILES
CCOP(Cl)(Cl)=O
Appearance (Color)
Clear colorless to light brown
Infrared spectrum
Conforms
Refractive index
1.4330 to 1.4350 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text