Thermo Scientific Chemicals

Ethyl 4-dimethylaminobenzoate, 99+%, Thermo Scientific Chemicals

Catalog number: 118401000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ethyl 4-dimethylaminobenzoate, 99+%, Thermo Scientific Chemicals

Catalog number: 118401000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 118401000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10287-53-3
IUPAC Name
ethyl 4-(dimethylamino)benzoate
Molecular Formula
C11H15NO2
InChI Key
FZUGPQWGEGAKET-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=C(C=C1)N(C)C
Appearance (Form)
Crystalline powder
Melting point
62°C to 65°C
Appearance (Color)
White to light yellow
GC
>=99.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text