Thermo Scientific Chemicals

Ethylene carbonate, 99+%, Thermo Scientific Chemicals

Catalog number: 118412500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethylene carbonate, 99+%, Thermo Scientific Chemicals

Catalog number: 118412500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118412500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
96-49-1
IUPAC Name
1,3-dioxolan-2-one
Molecular Formula
C3H4O3
InChI Key
KMTRUDSVKNLOMY-UHFFFAOYSA-N
SMILES
O=C1OCCO1
Infrared spectrum
Conforms
Appearance (Color)
White to yellow
Melting point
34°C to 37°C
Appearance (Form)
Low melting crystalline solid
GC
>=99.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Alkylation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text