Sample Holders
Sample Holders
Actual product may vary
Thermo Scientific™
Sample Holders
Get a more convenient, straightforward process with these Thermo Scientific™ Sample Holders, designed specifically for the Thermo Scientific™ ELEMENT GD™ GD-MS instrument.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1184170Rear Side Cooling, Thin Sample
1178090Sample Holder,Thin Sample
1190550Sample Holder, Standard
Catalog number 1184170
Price (USD)
-
Request A Quote
Type:
Rear Side Cooling, Thin Sample
Ask our AI about this Product
Get a more convenient, straightforward process with these Thermo Scientific™ Sample Holders, designed specifically for the Thermo Scientific™ ELEMENT GD™ GD-MS instrument. Even the most sophisticated elemental analysis, such as Glow Discharge Mass Spectrometry (GD-MS) is made more straightforward with the right accessories. These convenient Sample Holders are designed specifically for the ELEMENT GD GD-MS instrument, with options for standard or thin samples.
Gain Simplicity from Convenient Accessories
  • Sample holder featuring an ingenious clamping mechanism. For sample diameters up to 70mm, and maximum thickness of 40mm
  • Sample holder for thin samples, for use with rear side cooling; improves temperature stability of tiny samples. Maximum sample dimensions of 45 x 45mm and minimal thickness ∼0.1mm, depending on sample material
  • Rear-side cooling, for use with the sample holder for thin samples
Specifications
For Use With (Equipment)ELEMENT GD GD-MS Instrument
TypeRear Side Cooling, Thin Sample
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link
1x1 image pixel for data collection