Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, 99%, pure

Catalog number: 118432500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, 99%, pure

Catalog number: 118432500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 118432500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
60-00-4
IUPAC Name
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
Molecular Formula
C10H16N2O8
InChI Key
KCXVZYZYPLLWCC-UHFFFAOYSA-N
SMILES
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Titration Complexometric
>=98.5 %
Appearance (Form)
Crystalline powder
Melting point
237°C to 245°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text