Thermo Scientific Chemicals

Ethyl (ethoxymethylene)cyanoacetate, 98%, Thermo Scientific Chemicals

Catalog number: 118485000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl (ethoxymethylene)cyanoacetate, 98%, Thermo Scientific Chemicals

Catalog number: 118485000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118485000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94-05-3
IUPAC Name
ethyl (2Z)-2-cyano-3-ethoxyprop-2-enoate
Molecular Formula
C8H11NO3
InChI Key
KTMGNAIGXYODKQ-SREVYHEPSA-N
SMILES
CCO\C=C(\C#N)C(=O)OCC
GC
>=97.5 %
Melting point
47°C to 53°C
Appearance (Color)
White to yellow to yellow-brown
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals or flakes or crystalline solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text