Thermo Scientific Chemicals

2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 118520025
2.5 kg, Plastic drum, Each
Thermo Scientific Chemicals

2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 118520025
2.5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 118520025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
149-57-5
IUPAC Name
2-ethylhexanoic acid
Molecular Formula
C8H16O2
InChI Key
OBETXYAYXDNJHR-UHFFFAOYNA-N
SMILES
CCCCC(CC)C(O)=O
Color scale
=<15 APHA
GC
>=98.5 %
Water
=<0.1 % (K.F.)
Infrared spectrum
Conforms
Refractive index
1.4240 to 1.4260 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text