2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals
2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
500 g
1 kg
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 118520100
Price (USD)/ Each
203.65
Online exclusive
226.00 
Save 22.35 (10%)
-
Add to cart
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
203.65
Online exclusive
226.00 
Save 22.35 (10%)
Add to cart
2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals
Catalog number118520100
Price (USD)/ Each
203.65
Online exclusive
226.00 
Save 22.35 (10%)
-
Add to cart
Chemical Identifiers
CAS149-57-5
IUPAC Name2-ethylhexanoic acid
Molecular FormulaC8H16O2
InChI KeyOBETXYAYXDNJHR-UHFFFAOYNA-N
SMILESCCCCC(CC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<15 APHA
GC>=98.5 %
Water=<0.1 % (K.F.)
Infrared spectrumConforms
Refractive index1.4240 to 1.4260 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only