2-Ethyl-1-hexanol, 99%
2-Ethyl-1-hexanol, 99%
2-Ethyl-1-hexanol, 99%
2-Ethyl-1-hexanol, 99%
Thermo Scientific Chemicals

2-Ethyl-1-hexanol, 99%

CAS: 104-76-7 | C8H18O | 130.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 118530010
Price (USD)
47.90
Each
In stock
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
47.90
Each
Add to cart
Chemical Identifiers
CAS104-76-7
IUPAC Name2-ethylhexan-1-ol
Molecular FormulaC8H18O
InChI KeyYIWUKEYIRIRTPP-UHFFFAOYNA-N
SMILESCCCCC(CC)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=98.5 %
Refractive index1.4300 to 1.4330 (20°C, 589 nm)
Color scale=<10 APHA
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection