2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals
2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 118530025
Price (USD)/ Each
81.65
Online exclusive
90.30 
Save 8.65 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
81.65
Online exclusive
90.30 
Save 8.65 (10%)
Add to cart
2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals
Catalog number118530025
Price (USD)/ Each
81.65
Online exclusive
90.30 
Save 8.65 (10%)
-
Add to cart
Chemical Identifiers
CAS104-76-7
IUPAC Name2-ethylhexan-1-ol
Molecular FormulaC8H18O
InChI KeyYIWUKEYIRIRTPP-UHFFFAOYNA-N
SMILESCCCCC(CC)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=98.5 %
Color scale=<10 APHA
Water=<0.1 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only