Thermo Scientific Chemicals

2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals

Catalog number: 118530250
25 L, Metal drum, Each
Thermo Scientific Chemicals

2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals

Catalog number: 118530250
25 L, Metal drum, Each
Quantity
Packaging
Catalog number: 118530250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
104-76-7
IUPAC Name
2-ethylhexan-1-ol
Molecular Formula
C8H18O
InChI Key
YIWUKEYIRIRTPP-UHFFFAOYNA-N
SMILES
CCCCC(CC)CO
Appearance (Form)
Clear liquid
GC
>=98.5 %
Color scale
=<10 APHA
Water
=<0.1 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text