Thermo Scientific Chemicals

Ethyl 3-hydroxybutyrate, 99%, Thermo Scientific Chemicals

Catalog number: 118541000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 3-hydroxybutyrate, 99%, Thermo Scientific Chemicals

Catalog number: 118541000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118541000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5405-41-4
IUPAC Name
ethyl 3-hydroxybutanoate
Molecular Formula
C6H12O3
InChI Key
OMSUIQOIVADKIM-UHFFFAOYNA-N
SMILES
CCOC(=O)CC(C)O
Infrared spectrum
Conforms
Refractive index
1.4195 to 1.4215 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text