Ethyl 3-hydroxybutyrate, 99%, Thermo Scientific Chemicals
Ethyl 3-hydroxybutyrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3-hydroxybutyrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 118545000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5405-41-4
IUPAC Nameethyl 3-hydroxybutanoate
Molecular FormulaC6H12O3
InChI KeyOMSUIQOIVADKIM-UHFFFAOYNA-N
SMILESCCOC(=O)CC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4195 to 1.4215 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only