Ethyl L(-)-lactate, 97%, Thermo Scientific Chemicals
Ethyl L(-)-lactate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl L(-)-lactate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 118660010
Price (USD)/ Each
59.65
Online exclusive
69.70 
Save 10.05 (14%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
59.65
Online exclusive
69.70 
Save 10.05 (14%)
Add to cart
Ethyl L(-)-lactate, 97%, Thermo Scientific Chemicals
Catalog number118660010
Price (USD)/ Each
59.65
Online exclusive
69.70 
Save 10.05 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Nameethyl (2S)-2-hydroxypropanoate
Molecular FormulaC5H10O3
InChI KeyLZCLXQDLBQLTDK-BYPYZUCNSA-N
SMILESCCOC(=O)[C@H](C)O
Molecular Weight (g/mol)118.13
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
GC>=96.0 %
Refractive index1.4120 to 1.4140 (20°C, 589 nm)
Appearance (Form)Clear liquid
Water=<0.3 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only