Thermo Scientific Chemicals

Ethyl L(-)-lactate, 97%, Thermo Scientific Chemicals

Catalog number: 118660100
10 L, Plastic drum, Each
Thermo Scientific Chemicals

Ethyl L(-)-lactate, 97%, Thermo Scientific Chemicals

Catalog number: 118660100
10 L, Plastic drum, Each
Quantity
Packaging
Catalog number: 118660100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
687-47-8
IUPAC Name
ethyl (2S)-2-hydroxypropanoate
Molecular Formula
C5H10O3
InChI Key
LZCLXQDLBQLTDK-BYPYZUCNSA-N
SMILES
CCOC(=O)[C@H](C)O
Color scale
=<50 APHA
GC
>=96.0 %
Refractive index
1.4120 to 1.4140 (20°C, 589 nm)
Appearance (Form)
Clear liquid
Water
=<0.3 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text