Thermo Scientific Chemicals

Ethyl 2-mercaptoacetate, 99%, Thermo Scientific Chemicals

Catalog number: 118675000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl 2-mercaptoacetate, 99%, Thermo Scientific Chemicals

Catalog number: 118675000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118675000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
623-51-8
IUPAC Name
ethyl 2-sulfanylacetate
Molecular Formula
C4H8O2S
InChI Key
PVBRSNZAOAJRKO-UHFFFAOYSA-N
SMILES
CCOC(=O)CS
Appearance (Form)
Liquid
Refractive index
1.4560 to 1.4590 (20°C, 589 nm)
Appearance (Color)
Clear colorless
GC
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text