Thermo Scientific Chemicals

Ethylmercurithiosalicylic acid, sodium salt, 97.0-101.0%

Catalog number: 118680250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethylmercurithiosalicylic acid, sodium salt, 97.0-101.0%

Catalog number: 118680250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118680250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54-64-8
Appearance (Color)
White to cream
Infrared spectrum
Conforms
Additional info
ion mercuric: =< 0.7%
Loss on drying
=<0.5 % (vacuum/P2O5)
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Ethylmercurithiosalicylic acid is an oxidizing reagent that inhibits sulfhydryl-containing active sites of several enzymes
  • It is also known as thimerosal or 2-(ethylmercuriomercapto)benzoic acid sodium salt.
  • It can bind to sulfhydryl compounds, such as glutathione, cysteine, and sulfhydryl groups of proteins
  • Additionally, it can activate inositol triphosphate calcium channels on membranes of the endoplasmic reticulum, resulting in the regulation of cellular functions

Applications

  • Ethylmercurithiosalicylic acid has been widely investigated as a preservative in medicinal laboratory research, including vaccine studies
  • It has been shown to exhibit antifungal and antiseptic properties
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text