Thermo Scientific Chemicals

Ethyl oxalyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 118785000
500 mL, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl oxalyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: 118785000
500 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 118785000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4755-77-5
IUPAC Name
ethyl oxalochloridate
Molecular Formula
C4H5ClO3
InChI Key
OWZFULPEVHKEKS-UHFFFAOYSA-N
SMILES
CCOC(=O)C(Cl)=O
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.4160 to 1.4180 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text