Thermo Scientific Chemicals

Ethyl oxamate, 99%, Thermo Scientific Chemicals

Catalog number: 118795000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl oxamate, 99%, Thermo Scientific Chemicals

Catalog number: 118795000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 118795000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
617-36-7
IUPAC Name
ethyl carbamoylformate
Molecular Formula
C4H7NO3
InChI Key
RZMZBHSKPLVQCP-UHFFFAOYSA-N
SMILES
CCOC(=O)C(N)=O
Appearance (Color)
White
Infrared spectrum
Conforms
Assay
>=98.5 %
Appearance (Form)
Crystalline powder, crystals and/or chunks
Melting point
114°C to 119°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text