4-Ethylphenol, 97%
4-Ethylphenol, 97%
4-Ethylphenol, 97%
Thermo Scientific Chemicals

4-Ethylphenol, 97%

CAS: 123-07-9 | C8H10O | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
1 kg
Packaging:
Plastic bottle
Catalog number 118822500
Price (USD)
63.70
Each
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
63.70
Each
Add to cart
Chemical Identifiers
CAS123-07-9
IUPAC Name4-ethylphenol
Molecular FormulaC8H10O
InChI KeyHXDOZKJGKXYMEW-UHFFFAOYSA-N
SMILESCCC1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to brown
Infrared spectrumConforms
Appearance (Form)Crystals or crystalline solid or fused mass
GC>=96.0 %
Melting point38°C to 45°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link