Ethyl picolinate, 99%, Thermo Scientific Chemicals
Ethyl picolinate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl picolinate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 118860250
Price (USD)/ Each
62.60
-
Add to cart
Quantity:
25 mL
Request bulk or custom format
Price (USD)/ Each
62.60
Add to cart
Ethyl picolinate, 99%, Thermo Scientific Chemicals
Catalog number118860250
Price (USD)/ Each
62.60
-
Add to cart
Chemical Identifiers
CAS2524-52-9
IUPAC Nameethyl pyridine-2-carboxylate
Molecular FormulaC8H9NO2
InChI KeyFQYYIPZPELSLDK-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear yellow to pink
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.5090 to 1.5110 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only