Consumable Kits
Product Image
Thermo Scientific™
Consumable Kits
Everything needed for consistent HR-ICP-MS analysis with the Thermo Scientific™ ELEMENT™ Series analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1188700Consumable Element XR Glovebox
1188480Consumables Element XR Kit
1179480Consumables Element 2 Glovebox
1125671Consumables Element 2 Kit
1125664Spare Parts Kit
Catalog number 1188700
Price (USD)
-
Request A Quote
Type:
Consumable Element XR Glovebox
Ask our AI about this Product
Choose these convient, Thermo Scientific Consumable Kits of bundled spares, providing everything needed for consistent, highly advanced trace elemental analysis with your Thermo Scientific™ ELEMENT™ Series HR-ICP-MS system.
Select Quality Consumables for Improved Performance These useful and comprehensive kits are tailored to either the ELEMENT 2 or ELEMENT XR HR-ICP-MS analyzers to ensure you receive exactly what you need for your model of choice, from nebulizers and pump tubings, to cones, torch, and injector. even a spare Guard Electrode and load coil are included.
For the ELEMENT 2 HR-ICP-MS analyzer;
  • Consumables kit
  • Consumables kit with Glove Box
For the ELEMENT XR HR-ICP-MS analyzer;
  • Consumables kit
  • Consumables kit with Glove Box
For both the ELEMENT Series HR-ICP-MS instruments;
  • Spare parts kit, Hi-Pace Turbo-pump Version, includes replacement seals, valves, O-rings, reservoirs, oil, cones. For instruments shipped 2010 and later
Specifications
For Use With (Equipment)ELEMENT Series HR-ICP-MS system
IncludesNebulizers and Pump Tubings, to Cones, Torch, Injector, Spare Guard Electrode, and Load Coil
TypeConsumable Element XR Glovebox
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link