Thermo Scientific Chemicals

Ethyl N-piperazinecarboxylate, 99%, Thermo Scientific Chemicals

Catalog number: 118870250
25 g, Each
Thermo Scientific Chemicals

Ethyl N-piperazinecarboxylate, 99%, Thermo Scientific Chemicals

Catalog number: 118870250
25 g, Each
Quantity
Catalog number: 118870250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
120-43-4
IUPAC Name
ethyl piperazine-1-carboxylate
Molecular Formula
C7H14N2O2
InChI Key
LNOQURRKNJKKBU-UHFFFAOYSA-N
SMILES
CCOC(=O)N1CCNCC1
Refractive index
1.4760 to 1.4790 (20°C, 589 nm)
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Viscous liquid
Infrared spectrum
Conforms
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text