Ethyl pyruvate, 98%, Thermo Scientific Chemicals
Ethyl pyruvate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl pyruvate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 118941000
Price (USD)/ Each
81.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
96.40 
Save 14.75 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
81.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
96.40 
Save 14.75 (15%)
Add to cart
Ethyl pyruvate, 98%, Thermo Scientific Chemicals
Catalog number118941000
Price (USD)/ Each
81.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
96.40 
Save 14.75 (15%)
-
Add to cart
Chemical Identifiers
CAS617-35-6
IUPAC Nameethyl 2-oxopropanoate
Molecular FormulaC5H8O3
InChI KeyXXRCUYVCPSWGCC-UHFFFAOYSA-N
SMILESCCOC(=O)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.4040 to 1.4060 (20°C, 589 nm)
Appearance (Color)Clear light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only